Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3galt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020283.4 | NP_064679.2 | 2 | 1032 | ||
GenBank mRNA | AK035215.1 | - | 2 | 1032 | ||
BC094660.1 | - | 1 | 965 | 30543234 | ||
BC132246.1 | - | 1 | 887 | 40130523 | ||
BC132528.1 | - | 1 | 887 | 40130805 |