Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012001.2 | NP_036131.1 | 6 | 752 | ||
GenBank mRNA | AF071314.1 | - | 6 | 621 | ||
AK003752.1 | - | 6 | 614 | |||
AK009955.1 | - | 6 | 624 | |||
AK167833.1 | - | 6 | 638 | |||
BC020461.1 | - | 2 | 1174 | |||
BC116800.1 | - | 6 | 606 | 40126119 | ||
BC116826.1 | - | 6 | 606 | 40126145 | ||
Y17674.1 | - | 2 | 51 |