Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012001.2 | NP_036131.1 | 5 | 701 | ||
GenBank mRNA | AF071314.1 | - | 5 | 570 | ||
AK003752.1 | - | 5 | 563 | |||
AK009955.1 | - | 5 | 573 | |||
AK167833.1 | - | 5 | 587 | |||
BC020461.1 | - | 1 | 1123 | |||
BC116800.1 | - | 5 | 555 | 40126119 | ||
BC116826.1 | - | 5 | 555 | 40126145 |