Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Med14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048208.1 | NP_001041673.1 | 18 | 2615 | ||
NM_012005.3 | NP_036135.3 | 18 | 2615 | |||
GenBank mRNA | AK143808.1 | - | 4 | 557 | ||
BC065072.1 | - | 17 | 2801 | 6830820 |