Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012012.4 | NP_036142.2 | 11 | 1590 | ||
GenBank mRNA | AJ238213.1 | - | 9 | 1445 | ||
AK028728.1 | - | 11 | 1571 | |||
AK166425.1 | - | 11 | 1566 | |||
BC006671.1 | - | 11 | 1554 | 3499406 |