Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161712.1 | NP_001155184.1 | 2, 3 | 386 | ||
NM_013847.4 | NP_038875.3 | 2, 3 | 386 | |||
GenBank mRNA | AF093403.1 | - | 2, 3 | 344 | ||
AK007395.1 | - | 2, 3 | 306 | |||
AK013138.1 | - | 2, 3 | 341 | |||
AK075782.1 | - | 2, 3 | 326 | |||
AK076469.1 | - | 2, 3 | 359 | |||
AK087433.1 | - | 2, 3 | 345 | |||
AK183376.1 | - | 1, 2 | 161 | |||
AK187910.1 | - | 2, 3 | 162 | |||
BC024107.1 | - | 2, 3 | 386 | 5125361 |