Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012018.2 | NP_036148.2 | 28 | 4774 | ||
GenBank mRNA | AK088733.1 | - | 18 | 3137 | ||
AK131139.1 | - | 6 | 1128 | |||
BC079536.1 | - | 16 | 2799 | |||
BC158010.1 | - | 27 | 4647 | 9007318 |