Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012024.2 | NP_036154.1 | 3 | 869 | ||
GenBank mRNA | AK078894.1 | - | 3 | 899 | ||
AK088366.1 | - | 3 | 841 | |||
AK132432.1 | - | 3 | 902 | |||
AK133824.1 | - | 3 | 914 | |||
AK220163.1 | - | 3 | 859 | |||
BC085149.1 | - | 3 | 869 | 6848033 | ||
BC094387.1 | - | 3 | 861 | |||
U49728.1 | - | 1 | 146 |