Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012024.2 | NP_036154.1 | 3 | 911 | ||
GenBank mRNA | AK078894.1 | - | 3 | 941 | ||
AK088366.1 | - | 3 | 883 | |||
AK132432.1 | - | 3 | 944 | |||
AK133824.1 | - | 3 | 956 | |||
AK220163.1 | - | 3 | 901 | |||
BC085149.1 | - | 3 | 911 | 6848033 | ||
BC094387.1 | - | 3 | 903 | |||
U49728.1 | - | 1 | 188 |