Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253808.1 | NP_001240737.1 | 2 | 215 | ||
NM_001253809.1 | NP_001240738.1 | 2 | 211 | |||
NM_012025.7 | NP_036155.1 | 3 | 346 | |||
GenBank mRNA | AB030252.1 | - | 2 | 195 | ||
AF079974.1 | - | 2 | 156 | |||
AF212320.1 | - | 2 | 182 | |||
AF212321.1 | - | 3 | 313 | |||
AK053850.1 | - | 2 | 176 | |||
AK144608.1 | - | 3 | 314 | |||
AK144745.1 | - | 2 | 174 | |||
AK154929.1 | - | 3 | 334 | |||
AK162660.1 | - | 2 | 142 | |||
AK165897.1 | - | 2 | 168 | |||
AK166084.1 | - | 3 | 304 | |||
AK166094.1 | - | 2 | 173 | |||
AK168020.1 | - | 2 | 172 | |||
AK168025.1 | - | 2 | 172 | |||
AK168170.1 | - | 2 | 172 | |||
AK168679.1 | - | 2 | 156 | |||
BC010715.1 | - | 2 | 161 | 3602242 | ||
BY309801.1 | - | 2 | 215 |