Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mprip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012027.2 | NP_036157.2 | 15 | 2405 | ||
NM_201245.3 | NP_957697.1 | 15 | 2405 | |||
GenBank mRNA | AB093269.1 | - | 2 | 426 | ||
AK035435.1 | - | 9 | 1695 | |||
AK044512.1 | - | 14 | 2047 | |||
AK154807.1 | - | 16 | 2412 | |||
AK155630.1 | - | 15 | 2186 | |||
AK158296.1 | - | 15 | 2394 | |||
AK162530.1 | - | 13 | 1751 | |||
AK170849.1 | - | 15 | 2389 | |||
AK171089.1 | - | 15 | 2394 | |||
BC039932.1 | - | 14 | 1929 | |||
BC049803.1 | - | 15 | 1931 | |||
BC063067.1 | - | 15 | 2368 | 5721396 | ||
U73200.1 | - | 15 | 2265 |