Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr3e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164096.1 | NP_001157568.1 | 13, 14 | 1189 | ||
NM_025298.3 | NP_079574.2 | 14, 15 | 1267 | |||
GenBank mRNA | AK005034.1 | - | 13, 14 | 1190 | ||
AK012184.1 | - | 14, 15 | 1227 | |||
AK144721.1 | - | 14, 15 | 1231 | |||
AK146279.1 | - | 14, 15 | 1224 | |||
AK159723.1 | - | 14, 15 | 1224 | |||
BC028924.1 | - | 14, 15 | 1188 | |||
BC037637.1 | - | 14, 15 | 1217 | 5010681 |