Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012038.4 | NP_036168.1 | 3 | 588 | ||
GenBank mRNA | AJ489331.1 | - | 3 | 552 | ||
AK005091.2 | - | 2 | 150 | |||
AK144454.1 | - | 3 | 518 | |||
AK160224.1 | - | 3 | 520 | |||
AY101375.1 | - | 2 | 290 | |||
BC046226.1 | - | 3 | 493 | 5692284 | ||
D21165.1 | - | 2 | 290 | |||
EU373446.1 | - | 2 | 290 |