Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zw10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012039.2 | NP_036169.1 | 12 | 1744 | ||
GenBank mRNA | AF003951.1 | - | 2 | 80 | ||
AK145270.1 | - | 12 | 1713 | |||
AK167251.1 | - | 12 | 1713 | |||
AK167937.1 | - | 12 | 1673 | |||
AK168479.1 | - | 12 | 1713 | |||
BC012435.1 | - | 12 | 1693 | 3711269 | ||
BC025107.1 | - | 5 | 859 | |||
BC067405.1 | - | 3783 |