Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zw10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012039.2 | NP_036169.1 | 8, 9 | 1187 | ||
GenBank mRNA | AK167251.1 | - | 8, 9 | 1156 | ||
AK167937.1 | - | 8, 9 | 1116 | |||
AK168479.1 | - | 8, 9 | 1156 | |||
AK208616.1 | - | 2, 3 | 236 | |||
BC012435.1 | - | 8, 9 | 1136 | 3711269 | ||
BC067405.1 | - | 3226 |