Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cul1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012042.3 | NP_036172.1 | 9 | 1295 | ||
GenBank mRNA | AF083216.1 | - | 8 | 1019 | ||
AF136441.1 | - | 8 | 1150 | |||
AF176910.1 | - | 9 | 1184 | |||
AK147004.1 | - | 9 | 1277 | |||
AK164274.1 | - | 9 | 1262 | |||
AK165978.1 | - | 9 | 1236 | |||
AK209075.1 | - | 2 | 158 | |||
BC004836.1 | - | 8 | 1104 | |||
BC029260.1 | - | 9 | 1295 | 2812400 |