Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012044.2 | NP_036174.1 | 5 | 747 | ||
GenBank mRNA | AF112984.1 | - | 5 | 749 | ||
AF166098.1 | - | 5 | 743 | |||
BC027524.1 | - | 4 | 639 | 1533423 |