Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ermap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013848.2 | NP_038876.2 | 13 | 1579 | ||
GenBank mRNA | AF153906.1 | - | 13 | 1544 | ||
AK028170.1 | - | 2 | 318 | |||
AK036702.1 | - | 13 | 1540 | |||
AK158114.1 | - | 13 | 1558 | |||
BC063257.1 | - | 13 | 1513 | 6410069 |