Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012052.2 | NP_036182.1 | 6 | 719 | ||
GenBank mRNA | AK003181.1 | - | 6 | 657 | ||
AK010579.1 | - | 6 | 662 | |||
AK010678.1 | - | 6 | 660 | |||
AK011655.1 | - | 6 | 657 | |||
AK012234.1 | - | 6 | 661 | |||
AK012538.1 | - | 6 | 663 | |||
AK051243.1 | - | 6 | 662 | |||
AK143603.1 | - | 4 | 2699 | |||
AK146165.1 | - | 6 | 662 | |||
AK146796.1 | - | 6 | 662 | |||
AK148030.1 | - | 6 | 661 | |||
AK150479.1 | - | 6 | 655 | |||
AK159508.1 | - | 6 | 662 | |||
AK160526.1 | - | 6 | 661 | |||
AK164077.1 | - | 6 | 662 | |||
AK165174.1 | - | 6 | 651 | |||
AK166750.1 | - | 6 | 663 | |||
AK167210.1 | - | 6 | 661 | |||
AK167266.1 | - | 6 | 651 | |||
AK168845.1 | - | 6 | 651 | |||
AK197233.1 | - | 2 | 112 | |||
AK197426.1 | - | 2 | 114 | |||
AK201330.1 | - | 2 | 114 | |||
AK211385.1 | - | 2 | 114 | |||
BC010721.1 | - | 6 | 649 | 2811930 | ||
X76772.1 | - | 6 | 636 |