Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3d19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082414.2 | NP_001075883.2 | 15 | 2562 | ||
GenBank mRNA | AF003234.1 | - | 9 | 952 | ||
AK035963.1 | - | 1 | 1342 | |||
AK050185.1 | - | 7 | 819 | |||
BC028254.1 | - | 15 | 2110 | |||
BC031117.1 | - | 15 | 1673 | 5064197 | ||
D89677.2 | - | 21 | 3048 |