Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B9d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013717.2 | NP_038745.1 | 4 | 296 | ||
GenBank mRNA | AB030483.1 | - | 4 | 296 | ||
AK003038.1 | - | 4 | 291 | |||
AK010355.1 | - | 4 | 325 | |||
AK131701.1 | - | 4 | 287 | |||
BC008113.1 | - | 4 | 264 | 3483517 |