Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160229.1 | NP_001153701.1 | 5 | 572 | ||
NM_031494.2 | NP_113682.2 | 6 | 665 | |||
GenBank mRNA | AB221659.1 | - | 3 | 380 | ||
AK142957.1 | - | 5 | 572 | |||
BC138002.1 | - | 6 | 570 | 8861021 | ||
BC138003.1 | - | 6 | 570 | 8861022 | ||
BY010614.1 | - | 2 | 216 |