Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp275 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160229.1 | NP_001153701.1 | 5 | 865 | ||
NM_031494.2 | NP_113682.2 | 6 | 958 | |||
GenBank mRNA | AB221659.1 | - | 3 | 673 | ||
AK090015.1 | - | 1 | 124 | |||
AK142957.1 | - | 5 | 865 | |||
BC138002.1 | - | 6 | 863 | 8861021 | ||
BC138003.1 | - | 6 | 863 | 8861022 |