Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Podxl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013723.3 | NP_038751.2 | 8 | 1660 | ||
GenBank mRNA | AB028048.1 | - | 8 | 1541 | ||
AF290209.1 | - | 8 | 1559 | |||
AK134723.1 | - | 4 | 396 | |||
BC052442.1 | - | 8 | 1634 | 6406610 | ||
BC054530.1 | - | 8 | 1634 | 6406610 |