Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000525.3 | NP_000516.3 | 1 | 1879 | ||
NM_001166290.1 | NP_001159762.1 | 2 | 1224 | |||
XM_006718226.3 | XP_006718289.1 | 2 | 1508 | |||
XM_017017680.1 | XP_016873169.1 | 2 | 1104 | |||
GenBank mRNA | AK301550.1 | - | 2 | 1224 | ||
BC040617.1 | - | 2 | 1210 | |||
BC064497.1 | - | 1 | 1878 | 6279724 | ||
BC112358.1 | - | 1 | 1411 | 40032947 |