Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trp53bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290830.1 | NP_001277759.1 | 20 | 4441 | ||
NM_013735.4 | NP_038763.3 | 21 | 4591 | |||
GenBank mRNA | AJ414734.1 | - | 20 | 4434 | ||
AK029832.1 | - | 988 | ||||
AK036324.1 | - | 2 | 396 | |||
AK132323.1 | - | 3 | 624 | |||
BC035206.1 | - | 9 | 1803 | 3979746 | ||
BC079906.1 | - | 20 | 4375 | 30639375 |