Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159502.1 | NP_001152974.1 | 8 | 2541 | ||
NM_020494.3 | NP_065240.2 | 8 | 2375 | |||
GenBank mRNA | AF214732.1 | - | 8 | 2355 | ||
AK084251.1 | - | 1 | 959 | |||
AK149810.1 | - | 8 | 2328 | |||
AK150778.1 | - | 8 | 2327 | |||
AK152551.1 | - | 8 | 2327 | |||
AK152928.1 | - | 8 | 2327 | |||
AK166360.1 | - | 8 | 2336 | |||
BC043721.1 | - | 8 | 2343 | |||
BC055048.1 | - | 8 | 2305 | 6484820 | ||
BC055317.1 | - | 8 | 2541 | 5696084 | ||
U46690.1 | - | 6 | 1149 |