Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nufip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013745.5 | NP_038773.1 | 7 | 860 | ||
GenBank mRNA | AF159549.1 | - | 7 | 818 | ||
AK032324.1 | - | 7 | 861 | |||
AK156205.1 | - | 7 | 847 | |||
BC020364.1 | - | 7 | 835 | |||
BC056192.1 | - | 7 | 846 | 30024494 |