Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163182.1 | NP_001156654.1 | 8 | 807 | ||
NM_001163183.1 | NP_001156655.1 | 8 | 765 | |||
NM_001163184.1 | NP_001156656.1 | 7 | 660 | |||
NM_001163185.1 | NP_001156657.1 | 7 | 650 | |||
NM_001163186.1 | NP_001156658.1 | 7 | 756 | |||
NM_001163187.1 | NP_001156659.1 | 6 | 651 | |||
NM_001291336.1 | NP_001278265.1 | 6 | 620 | |||
NM_013746.3 | NP_038774.1 | 9 | 912 | |||
GenBank mRNA | AF000272.1 | - | 9 | 883 | ||
AF071000.1 | - | 6 | 639 | |||
AF100613.1 | - | 7 | 744 | |||
AF101053.1 | - | 8 | 778 | |||
AK005102.1 | - | 7 | 615 | |||
AK141634.1 | - | 618 | ||||
AK158172.1 | - | 7 | 651 | |||
AK164683.1 | - | 7 | 615 | |||
BC024756.1 | - | 9 | 873 | 5369461 | ||
CX201870.1 | - | 8 | 717 |