Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phlda3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013750.2 | NP_038778.1 | 2 | 1104 | ||
GenBank mRNA | AK003767.1 | - | 2 | 1070 | ||
AK218246.1 | - | 1 | 198 | |||
BC023376.1 | - | 2 | 1013 | |||
BC023408.1 | - | 2 | 1013 | 5252055 |