Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163311.1 | NP_001156783.1 | 4 | 560 | ||
NM_013761.4 | NP_038789.1 | 5 | 786 | |||
GenBank mRNA | AF148321.1 | - | 4 | 441 | ||
AK017235.1 | - | 5 | 665 | |||
AK028034.1 | - | 4 | 443 | |||
AK031687.1 | - | 5 | 658 | |||
AK043738.1 | - | 5 | 692 | |||
AK043819.1 | - | 4 | 466 | |||
AK080830.1 | - | 5 | 666 | |||
AK157122.1 | - | 4 | 711 | |||
AK170096.1 | - | 5 | 672 | |||
AK170471.1 | - | 5 | 672 | |||
BC011164.1 | - | 5 | 663 | 4195695 |