Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txnl4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038608.2 | NP_001033697.2 | 2 | 268 | ||
NM_001042408.1 | NP_001035867.1 | 2 | 244 | |||
NM_025299.3 | NP_079575.1 | 2 | 268 | |||
NM_178604.3 | NP_848719.2 | 2 | 380 | |||
GenBank mRNA | AK004039.1 | - | 2 | 225 | ||
AK007331.1 | - | 2 | 230 | |||
AK010599.1 | - | 2 | 268 | |||
AK011717.1 | - | 2 | 332 | |||
AK148287.1 | - | 2 | 251 | |||
AK166497.1 | - | 2 | 244 | |||
AK171121.1 | - | 2 | 220 | |||
AK181260.1 | - | 1 | 58 | |||
AK184254.1 | - | 1 | 58 | |||
AK189069.1 | - | 1 | 59 | |||
AK189366.1 | - | 1 | 58 | |||
AK198184.1 | - | 1 | 58 | |||
AK218707.1 | - | 1 | 58 | |||
BC003876.1 | - | 2 | 196 | |||
BC014770.1 | - | 2 | 244 | |||
BC025053.1 | - | 2 | 221 | |||
BC029327.1 | - | 2 | 238 | |||
BC031634.1 | - | 2 | 209 | 3662554 | ||
BG086914.2 | - | 1 | 157 | |||
BU514237.1 | - | 2 | 181 | |||
BY065764.1 | - | 2 | 269 |