Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025571.2 | NP_001020742.1 | 7 | 1372 | ||
NM_021309.3 | NP_067284.1 | 7 | 1396 | |||
GenBank mRNA | AF203343.1 | - | 7 | 1403 | ||
AK037489.1 | - | 7 | 1094 | |||
AK172182.1 | - | 7 | 1124 | |||
BC034847.1 | - | 7 | 1104 | 3327113 | ||
U69490.1 | - | 7 | 1082 |