Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163218.1 | NP_001156690.1 | 5 | 589 | ||
NM_013766.2 | NP_038794.1 | 5 | 559 | |||
GenBank mRNA | AB019118.1 | - | 5 | 556 | ||
AF150740.1 | - | 5 | 569 | |||
AF234637.1 | - | 5 | 559 | |||
AK014980.1 | - | 5 | 589 | |||
BC120576.1 | - | 5 | 547 | 40129499 | ||
BC120578.1 | - | 5 | 547 | 40129501 |