Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013348.3 | NP_037480.1 | 2 | 364 | ||
GenBank mRNA | AF081466.1 | - | 2 | 562 | ||
BC035918.1 | - | 1 | 652 | 5531571 |