Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Magel2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013779.2 | NP_038807.4 | 1 | 3729 | ||
GenBank mRNA | AJ243608.1 | - | 1 | 1244 | ||
AK082944.1 | - | 1 | 3702 | |||
AK086725.1 | - | 1 | 3724 | |||
BC054763.1 | - | 1 | 1021 | |||
BC062913.1 | - | 3 | 3234 | |||
BC071222.1 | - | 1 | 1507 |