Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dusp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007268.1 | NP_001007269.1 | 7 | 1114 | ||
NM_013849.3 | NP_038877.2 | 3 | 439 | |||
GenBank mRNA | AB027003.1 | - | 2 | 220 | ||
AF237620.1 | - | 3 | 422 | |||
AK006247.1 | - | 3 | 440 | |||
AY674052.1 | - | 7 | 1114 | |||
BC100421.1 | - | 2 | 345 | 30919819 |