Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017404.4 | NP_059100.3 | 6 | 678 | ||
GenBank mRNA | AF239728.1 | - | 6 | 595 | ||
AK007681.1 | - | 6 | 614 | |||
AK031942.1 | - | 6 | 599 | |||
BC012274.1 | - | 6 | 557 | 3981658 | ||
BC016561.1 | - | 6 | 614 | 2645316 | ||
BC036276.1 | - | 6 | 614 |