Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177658.1 | NP_001171129.1 | 3 | 360 | ||
NM_025300.4 | NP_079576.1 | 3 | 360 | |||
GenBank mRNA | AK003026.1 | - | 3 | 341 | ||
AK003367.1 | - | 3 | 344 | |||
AK010474.1 | - | 3 | 320 | |||
AK010604.1 | - | 3 | 353 | |||
AK011775.1 | - | 2 | 180 | |||
AK017804.1 | - | 3 | 337 | |||
AK017820.1 | - | 3 | 339 | |||
AK017824.1 | - | 3 | 323 | |||
AK019175.1 | - | 3 | 333 | |||
AK019244.1 | - | 3 | 320 | |||
AK019249.1 | - | 3 | 320 | |||
AK150880.1 | - | 3 | 361 | |||
AK160660.1 | - | 3 | 318 | |||
BC021304.1 | - | 3 | 356 | 3498041 | ||
BC027233.1 | - | 3 | 345 | 3498065 | ||
BC068230.1 | - | 3 | 343 |