Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270422.1 | NP_001257351.1 | 6 | 1144 | ||
NM_001291622.1 | NP_001278551.1 | 6 | 1098 | |||
NM_001291623.1 | NP_001278552.1 | 3 | 891 | |||
NM_001291624.1 | NP_001278553.1 | 4 | 1019 | |||
NM_001291625.1 | NP_001278554.1 | 5 | 1100 | |||
NM_018658.2 | NP_061128.2 | 5 | 1016 | |||
NM_170741.2 | NP_733937.2 | 4 | 935 | |||
NM_170742.2 | NP_733938.2 | 4 | 972 | |||
XM_005257337.4 | XP_005257394.1 | 3 | 818 | |||
XM_006721885.3 | XP_006721948.1 | 7 | 1224 | |||
XM_006721886.3 | XP_006721949.1 | 6 | 1096 | |||
XM_006721887.3 | XP_006721950.1 | 6 | 1089 | |||
XM_011524781.2 | XP_011523083.1 | 5 | 1055 | |||
XM_017024609.1 | XP_016880098.1 | 7 | 1327 | |||
XM_017024610.1 | XP_016880099.1 | 6 | 1062 | |||
XM_017024611.1 | XP_016880100.1 | 6 | 1136 | |||
XM_017024612.1 | XP_016880101.1 | 5 | 1055 | |||
XM_017024613.1 | XP_016880102.1 | 6 | 1183 | |||
GenBank mRNA | AF153815.1 | - | 5 | 956 | ||
AF153816.1 | - | 5 | 956 | |||
AF153817.1 | - | 3 | 728 | |||
AF179353.1 | - | 1 | 470 | |||
AK225944.1 | - | 8 | 981 | |||
AK290799.1 | - | 4 | 825 | |||
BC033038.2 | - | 4 | 962 | 5262150 |