Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285980.1 | NP_001272909.1 | 6 | 890 | ||
NM_013787.3 | NP_038815.1 | 6 | 960 | |||
GenBank mRNA | AF083215.1 | - | 6 | 823 | ||
AK006221.1 | - | 6 | 890 | |||
AK032979.1 | - | 6 | 871 | |||
AK044212.1 | - | 6 | 861 | |||
AK166034.1 | - | 6 | 861 | |||
AK167677.1 | - | 6 | 858 | |||
BC092236.1 | - | 6 | 834 | 30610417 | ||
BC098371.1 | - | 6 | 943 |