Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abca8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013851.2 | NP_038879.2 | 34 | 4523 | ||
GenBank mRNA | AF213393.1 | - | 1 | 16 | ||
AF498362.1 | - | 34 | 4523 | |||
AK043744.1 | - | 72 | ||||
AK173034.1 | - | 33 | 4264 | |||
BC089557.1 | - | 10 | 1166 | |||
BC141401.1 | - | 34 | 4486 | 9056014 |