Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030239.2 | NP_084515.2 | 6 | 753 | ||
GenBank mRNA | AF324242.1 | - | 6 | 705 | ||
AK030831.1 | - | 6 | 755 | |||
AK051880.1 | - | 6 | 719 | |||
AK156626.1 | - | 6 | 721 | |||
BC029095.1 | - | 5 | 598 | |||
BC138772.1 | - | 6 | 734 | 8861794 | ||
BC138773.1 | - | 6 | 733 | 8861795 |