Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013852.2 | NP_038880.1 | 9 | 1016 | ||
GenBank mRNA | AK030626.1 | - | 9 | 981 | ||
AK179781.1 | - | 3 | 246 | |||
AK181390.1 | - | 1 | 12 | |||
AK211939.1 | - | 1 | 12 | |||
BC032923.1 | - | 9 | 957 | 1430389 |