Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021022.3 | NP_066302.2 | 2 | 183 | ||
GenBank mRNA | AF133903.1 | - | 2 | 154 | ||
AI316041.1 | - | 2 | 180 | |||
AK144553.1 | - | 3 | 260 |