Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sergef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013789.2 | NP_038817.1 | 11 | 1365 | ||
GenBank mRNA | AJ243952.1 | - | 11 | 1348 | ||
AK145819.1 | - | 11 | 1354 | |||
BC049603.1 | - | 11 | 1344 | 6591072 | ||
BC119533.1 | - | 11 | 1348 | 40056360 | ||
BC119534.1 | - | 11 | 1348 | 40056367 |