Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319945.1 | NP_001306874.1 | 1 | 348 | ||
NM_138387.3 | NP_612396.1 | 1 | 348 | |||
NR_028581.1 | 1 | 348 | ||||
NR_028582.1 | 1 | 348 | ||||
XM_011525474.2 | XP_011523776.1 | 1 | 328 | |||
GenBank mRNA | BC002494.2 | - | 1 | 146 | 3050476 | |
BC021574.1 | - | 1 | 328 | 4842511 | ||
BM790097.1 | - | 1 | 343 | |||
DB460416.1 | - | 1 | 348 |