Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013794.4 | NP_038822.3 | 6 | 735 | ||
GenBank mRNA | AF074458.1 | - | 6 | 785 | ||
AF074461.1 | - | 4 | 442 | |||
AF074462.1 | - | 3 | 353 | |||
AF146570.4 | - | 6 | 735 | |||
AF218080.1 | - | 5 | 654 | |||
AF218081.1 | - | 4 | 455 | |||
AF218082.1 | - | 3 | 365 | |||
AY971807.1 | - | 5 | 633 | |||
BC119242.1 | - | 6 | 724 | 8733991 | ||
BC144924.1 | - | 6 | 724 | 9053472 | ||
BC144940.1 | - | 6 | 762 | 9053488 |