Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013794.4 | NP_038822.3 | 4 | 416 | ||
GenBank mRNA | AF074458.1 | - | 4 | 466 | ||
AF146570.4 | - | 4 | 416 | |||
AF218080.1 | - | 3 | 335 | |||
AY971807.1 | - | 3 | 314 | |||
BC119242.1 | - | 4 | 405 | 8733991 | ||
BC144924.1 | - | 4 | 405 | 9053472 | ||
BC144940.1 | - | 4 | 443 | 9053488 |
Gene Symbol: Klra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK172530.1 | - | 367 |