Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nagpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013796.3 | NP_038824.2 | 3, 4 | 719 | ||
GenBank mRNA | AK032158.1 | - | 3, 4 | 720 | ||
AK138035.1 | - | 3, 4 | 712 | |||
AK155450.1 | - | 3, 4 | 698 | |||
AK159629.1 | - | 3, 4 | 698 | |||
AK162633.1 | - | 3, 4 | 701 | |||
AK168830.1 | - | 2, 3 | 806 | |||
AK170219.1 | - | 3, 4 | 701 | |||
BC019640.1 | - | 2, 3 | 781 | |||
BC039790.1 | - | 3, 4 | 711 | 4951805 |